The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108248.1 | NP_001101718.1 | 11 - 12 | 1115 | 82 |
GenBank mRNA | BC158677.1 | - | 11 - 12 | 1502 | 82 |