The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004206.1 | NP_001004206.1 | 12 - 13 | 1238 | 78 |
GenBank mRNA | BC079095.1 | - | 12 - 13 | 1238 | 78 |