Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bgn
Entrez Gene ID
25181
Gene Name
biglycan
Gene Aliases
BSPG1
Chromosome Location
Chr.X: 157319042 - 157331204 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017087.1 NP_058783.1 4 - 5 690 70
GenBank mRNA AY724513.1 - 4 - 5 693 70
BC072480.1 - 4 - 5 698 70
U17834.1 - 4 - 5 690 70

View Full Product Details