The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135839.1 | NP_001129311.1 | 6 - 7 | 528 | 82 |
XM_006229591.2 | XP_006229653.1 | 6 - 7 | 838 | 82 | |
XM_006229592.2 | XP_006229654.1 | 6 - 7 | 637 | 82 | |
GenBank mRNA | FQ235221.1 | - | 6 - 7 | 472 | 82 |