Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btn1a1
Entrez Gene ID
306956
Gene Name
butyrophilin, subfamily 1, member A1
Gene Aliases
-
Chromosome Location
Chr.17: 43899565 - 43908986 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170337.1 NP_001163808.1 4 - 5 859 75
XM_006253970.2 XP_006254032.1 5 - 6 1238 75
GenBank mRNA AY327120.1 - 4 - 5 863 75

View Full Product Details