The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034947.1 | NP_001030119.1 | 6 - 7 | 894 | 149 |
GenBank mRNA | BC101920.1 | - | 6 - 7 | 894 | 149 |