The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020097.1 | NP_064482.1 | 1 - 2 | 2156 | 81 |
XM_006252206.2 | XP_006252268.1 | 4 - 5 | 3462 | 81 | |
GenBank mRNA | AB033367.1 | - | 1 - 2 | 2156 | 81 |