Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc25
Entrez Gene ID
361059
Gene Name
coiled-coil domain containing 25
Gene Aliases
RGD1307689
Chromosome Location
Chr.15: 42522108 - 42555318 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108382.1 NP_001101852.1 8 - 9 704 115
XM_006252132.2 XP_006252194.1 8 - 9 703 115

View Full Product Details