Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp57
Entrez Gene ID
361783
Gene Name
zinc finger protein 57
Gene Aliases
-
Chromosome Location
Chr.20: 2014177 - 2020488 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213565.4 NP_998730.2 2 - 3 168 67
GenBank mRNA CA511208.1 - 2 - 3 169 67
DY312591.1 - 2 - 3 220 67

View Full Product Details