Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp57
Entrez Gene ID
361783
Gene Name
zinc finger protein 57
Gene Aliases
-
Chromosome Location
Chr.20: 2014177 - 2020488 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213565.4 NP_998730.2 3 - 4 289 72
XM_008772694.1 XP_008770916.1 2 - 3 462 72
GenBank mRNA BC127468.1 - 1 - 2 107 72
CA511208.1 - 3 - 4 290 72
DY312591.1 - 3 - 4 341 72

View Full Product Details