Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim26
Entrez Gene ID
309586
Gene Name
tripartite motif-containing 26
Gene Aliases
Znf173
Chromosome Location
Chr.20: 2203176 - 2224463 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772692.1 XP_008770914.1 6 - 7 946 69

View Full Product Details