The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106376.1 | NP_001099846.1 | 7 - 8 | 1227 | 74 |
XM_008772691.1 | XP_008770913.1 | 8 - 9 | 1138 | 74 |