Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bex1
Entrez Gene ID
501625
Gene Name
brain expressed, X-linked 1
Gene Aliases
-
Chromosome Location
Chr.X: 106761388 - 106771656 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY208292.1 - - 47 67

View Full Product Details