Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hmgxb4
Entrez Gene ID
307667
Gene Name
HMG box domain containing 4
Gene Aliases
Hmg2l1, Hmgb2l1
Chromosome Location
Chr.19: 14352673 - 14390499 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107412.1 NP_001100882.1 10 - 11 1877 58

View Full Product Details