Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hmgxb4
Entrez Gene ID
307667
Gene Name
HMG box domain containing 4
Gene Aliases
Hmg2l1, Hmgb2l1
Chromosome Location
Chr.19: 14352673 - 14390499 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107412.1 NP_001100882.1 9 - 10 1756 92
XM_006255141.2 XP_006255203.1 8 - 9 6174 92
XM_006255143.1 XP_006255205.1 9 - 10 1758 92
XM_006255144.1 XP_006255206.1 7 - 8 1563 92
XM_008772312.1 XP_008770534.1 7 - 8 1741 92

View Full Product Details