Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npc1
Entrez Gene ID
266732
Gene Name
Niemann-Pick disease, type C1
Gene Aliases
Cdig2
Chromosome Location
Chr.18: 3616878 - 3662715 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153624.2 NP_705888.2 23 - 24 3678 84
XM_006254403.2 XP_006254465.1 23 - 24 3734 84
GenBank mRNA BC166779.1 - 5 - 6 716 84

View Full Product Details