The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012223.1 | NP_001012223.1 | 2 - 3 | 204 | 87 |
XM_008765773.1 | XP_008763995.1 | 2 - 3 | 231 | 87 | |
GenBank mRNA | BC079406.1 | - | 2 - 3 | 204 | 87 |