Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt7
Entrez Gene ID
300242
Gene Name
keratin 7, type II
Gene Aliases
Krt2-7
Chromosome Location
Chr.7: 143059734 - 143075907 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750407.3 XP_003750455.1 4 - 5 1077 62

View Full Product Details