Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scaf11
Entrez Gene ID
312030
Gene Name
SR-related CTD-associated factor 11
Gene Aliases
Sfrs2ip, Srsf2ip
Chromosome Location
Chr.7: 137804019 - 137856337 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271170.1 NP_001258099.1 4 - 5 360 148
GenBank mRNA CB547108.1 - 4 - 5 366 148

View Full Product Details