Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bcas2
Entrez Gene ID
295334
Gene Name
breast carcinoma amplified sequence 2
Gene Aliases
-
Chromosome Location
Chr.2: 205662633 - 205670514 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106458.2 NP_001099928.1 2 - 3 200 99
GenBank mRNA BC169122.1 - 2 - 3 200 99
FQ226799.1 - 2 - 3 193 99

View Full Product Details