Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Capza1
Entrez Gene ID
691149
Gene Name
capping protein (actin filament) muscle Z-line, alpha 1
Gene Aliases
-
Chromosome Location
Chr.2: 207305748 - 207350812 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109625.2 NP_001103095.2 4 - 5 507 84
GenBank mRNA BC166464.1 - 4 - 5 222 84
EV765154.1 - 4 - 5 507 84

View Full Product Details