Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnd3
Entrez Gene ID
65195
Gene Name
potassium channel, voltage-gated Shal-related subfamily D, member 3
Gene Aliases
Kv4.3
Chromosome Location
Chr.2: 207923775 - 208140727 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270962.1 NP_001257891.1 6 - 7 1609 101
GenBank mRNA AB003587.1 - 5 - 6 1517 101
AF334791.1 - 5 - 6 1517 101
U92897.1 - 2 - 3 155 101

View Full Product Details