Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vtcn1
Entrez Gene ID
295322
Gene Name
V-set domain containing T cell activation inhibitor 1
Gene Aliases
RGD1311204
Chromosome Location
Chr.2: 203200435 - 203274264 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024244.1 NP_001019415.1 5 - 6 982 133
GenBank mRNA BC095842.1 - 5 - 6 982 133

View Full Product Details