Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC103689947
Entrez Gene ID
103689947
Gene Name
selenium-binding protein 1
Gene Aliases
-
Chromosome Location
Chr.2: 195416472 - 195423665 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008761279.1 XP_008759501.1 - 541 116

Gene Symbol
Selenbp1
Entrez Gene ID
140927
Gene Name
selenium binding protein 1
Gene Aliases
Selenbp2
Chromosome Location
Chr.2: 196105592 - 196146474 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080892.1 NP_543168.1 4 - 5 541 116
GenBank mRNA AB036799.1 - 4 - 5 541 116
BC074008.1 - 4 - 5 551 116
FQ219337.1 - 4 - 5 521 116

View Full Product Details