Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aadat
Entrez Gene ID
29416
Gene Name
aminoadipate aminotransferase
Gene Aliases
Kat2
Chromosome Location
Chr.16: 32832001 - 32868680 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017193.1 NP_058889.1 6 - 7 831 135
XM_006253067.2 XP_006253129.1 6 - 7 919 135
GenBank mRNA BC078864.1 - 6 - 7 827 135
Z50144.1 - 6 - 7 831 135

View Full Product Details