Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cfd
Entrez Gene ID
54249
Gene Name
complement factor D (adipsin)
Gene Aliases
Adn, Df, EVE
Chromosome Location
Chr.7: 12634216 - 12635939 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077642.1 NP_001071110.1 2 - 3 212 52
GenBank mRNA M92059.1 - 2 - 3 209 52
S73894.1 - 2 - 3 212 52

View Full Product Details