Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Elane
Entrez Gene ID
299606
Gene Name
elastase, neutrophil expressed
Gene Aliases
Ela2
Chromosome Location
Chr.7: 12638320 - 12640168 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106767.1 NP_001100237.1 1 - 2 89 91

View Full Product Details