Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Itch
Entrez Gene ID
311567
Gene Name
itchy E3 ubiquitin protein ligase
Gene Aliases
-
Chromosome Location
Chr.3: 150660724 - 150752016 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005887.1 NP_001005887.1 5 - 6 493 146
XM_008762335.1 XP_008760557.1 7 - 8 721 146
XM_008762336.1 XP_008760558.1 7 - 8 646 146
XM_008762337.1 XP_008760559.1 4 - 5 398 146
GenBank mRNA AY600518.1 - 5 - 6 493 146

View Full Product Details