Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tepp
Entrez Gene ID
291850
Gene Name
testis, prostate and placenta expressed
Gene Aliases
-
Chromosome Location
Chr.19: 10127607 - 10138436 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201655.1 NP_970617.1 7 - 8 804 100
XM_008772274.1 XP_008770496.1 6 - 7 1187 100

View Full Product Details