Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ces5a
Entrez Gene ID
307660
Gene Name
carboxylesterase 5A
Gene Aliases
Ces5, Ces7
Chromosome Location
Chr.19: 12097529 - 12136579 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012056.1 NP_001012056.1 12 - 13 1612 63
GenBank mRNA AF479659.1 - 12 - 13 1612 63

View Full Product Details