Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cklf
Entrez Gene ID
245978
Gene Name
chemokine-like factor
Gene Aliases
Cklf1
Chromosome Location
Chr.19: 904758 - 914880 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF253064.2 - 1 - 2 243 93

View Full Product Details