Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Coq9
Entrez Gene ID
498909
Gene Name
coenzyme Q9
Gene Aliases
-
Chromosome Location
Chr.19: 10583855 - 10596848 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035257.1 NP_001030334.1 7 - 8 862 90
GenBank mRNA BC079370.1 - 7 - 8 848 90
BC104702.1 - 7 - 8 848 90
BC127449.1 - 7 - 8 863 90

View Full Product Details