The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108865.1 | NP_001102335.1 | 3 - 4 | 673 | 132 |
GenBank mRNA | AB002591.1 | - | 3 - 4 | 924 | 132 |
FQ229094.1 | - | 3 - 4 | 655 | 132 |