Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Irx4
Entrez Gene ID
306655
Gene Name
iroquois homeobox 4
Gene Aliases
-
Chromosome Location
Chr.1: 32634774 - 32645157 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107330.1 NP_001100800.1 1 - 2 367 53

View Full Product Details