The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107333.1 | NP_001100803.1 | 10 - 11 | 1315 | 119 |
XM_006227781.2 | XP_006227843.1 | 10 - 11 | 5065 | 119 | |
XM_006227782.2 | XP_006227844.1 | 10 - 11 | 1702 | 119 | |
XM_008758690.1 | XP_008756912.1 | 10 - 11 | 1432 | 119 |