Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc2a12
Entrez Gene ID
308028
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 12
Gene Aliases
-
Chromosome Location
Chr.1: 24003625 - 24056373 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107451.1 NP_001100921.1 4 - 5 1890 133

View Full Product Details