Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep72
Entrez Gene ID
308064
Gene Name
centrosomal protein 72
Gene Aliases
RGD1566336
Chromosome Location
Chr.1: 31834977 - 31864938 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008758697.1 XP_008756919.1 7 - 8 1148 135
XM_217737.8 XP_217737.4 7 - 8 1254 135

View Full Product Details