Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep72
Entrez Gene ID
308064
Gene Name
centrosomal protein 72
Gene Aliases
RGD1566336
Chromosome Location
Chr.1: 31834977 - 31864938 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006227800.2 XP_006227862.1 8 - 9 1214 76
XM_006227801.2 XP_006227863.1 8 - 9 1151 76
XM_008758697.1 XP_008756919.1 8 - 9 1287 76
XM_217737.8 XP_217737.4 8 - 9 1393 76

View Full Product Details