The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138831.1 | NP_620186.1 | 5 - 6 | 1501 | 153 |
XM_008772974.1 | XP_008771196.1 | 5 - 6 | 1540 | 153 | |
XM_008772975.1 | XP_008771197.1 | 4 - 5 | 1395 | 153 | |
GenBank mRNA | AB047324.1 | - | 5 - 6 | 1501 | 153 |