Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csn1s2b
Entrez Gene ID
286759
Gene Name
casein alpha s2-like B
Gene Aliases
Csnd
Chromosome Location
Chr.14: 21899954 - 21916068 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173106.1 NP_775129.1 - 173 124
GenBank mRNA AY154894.1 - - 173 124

View Full Product Details