Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pafah2
Entrez Gene ID
313611
Gene Name
platelet-activating factor acetylhydrolase 2
Gene Aliases
-
Chromosome Location
Chr.5: 152606853 - 152635956 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177932.2 NP_808793.1 6 - 7 673 78
XM_006239127.2 XP_006239189.1 7 - 8 974 78
XM_008764144.1 XP_008762366.1 7 - 8 976 78
GenBank mRNA AY225592.1 - 6 - 7 673 78

View Full Product Details