Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stmn2
Entrez Gene ID
84510
Gene Name
stathmin 2
Gene Aliases
Scg10, Scgn10
Chromosome Location
Chr.2: 95424251 - 95471900 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053440.2 NP_445892.1 1 - 2 38 85
GenBank mRNA AF306458.1 - 1 - 2 136 85
BC087660.1 - 1 - 2 38 85

View Full Product Details