Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcas1
Entrez Gene ID
246755
Gene Name
breast carcinoma amplified sequence 1
Gene Aliases
Band83, Pmes-2
Chromosome Location
Chr.3: 167957994 - 168033457 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB029897.1 - 6 - 7 824 106
AB180272.1 - 5 - 6 689 106

View Full Product Details