Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gast
Entrez Gene ID
25320
Gene Name
gastrin
Gene Aliases
Gas, PPG34
Chromosome Location
Chr.10: 88245532 - 88248484 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012849.1 NP_036981.1 1 - 2 41 60
GenBank mRNA M38653.1 - 1 - 2 41 60

View Full Product Details