Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cgref1
Entrez Gene ID
245918
Gene Name
cell growth regulator with EF hand domain 1
Gene Aliases
Cgr11
Chromosome Location
Chr.6: 26797126 - 26809132 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139087.2 NP_620787.2 1 - 2 127 87
GenBank mRNA FM040765.1 - 1 - 2 127 87
FQ220335.1 - 1 - 2 57 87
U66470.1 - 1 - 2 48 86

View Full Product Details