The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037310.1 | NP_001032387.1 | 3 - 3 | 1153 | 135 |
NM_184048.2 | NP_908937.2 | 3 - 3 | 674 | 135 | |
GenBank mRNA | AY325235.1 | - | 2 - 2 | 510 | 135 |
AY359819.1 | - | 1 - 1 | 470 | 135 | |
BC091109.1 | - | 3 - 3 | 663 | 135 | |
FQ213120.1 | - | 3 - 3 | 657 | 135 | |
FQ215462.1 | - | 3 - 3 | 668 | 135 | |
FQ229736.1 | - | 3 - 3 | 667 | 135 |