The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199237.1 | NP_954707.1 | 9 - 10 | 921 | 113 |
XM_008764476.1 | XP_008762698.1 | 7 - 8 | 581 | 113 | |
GenBank mRNA | L02842.1 | - | 9 - 10 | 921 | 113 |