Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grm5
Entrez Gene ID
24418
Gene Name
glutamate receptor, metabotropic 5
Gene Aliases
mGluR5, mGlur5
Chromosome Location
Chr.1: 151207859 - 151784991 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017012.1 NP_058708.1 4 - 5 1407 160
XM_006229659.2 XP_006229721.1 4 - 5 1734 160
GenBank mRNA D10891.1 - 4 - 5 1407 160
EU559292.1 - 3 - 4 1140 160

View Full Product Details