Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Harbi1
Entrez Gene ID
690164
Gene Name
harbinger transposase derived 1
Gene Aliases
-
Chromosome Location
Chr.3: 80614882 - 80624168 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113793.3 NP_001107265.2 1 - 2 34 120
GenBank mRNA BC158734.1 - 1 - 2 34 120

View Full Product Details