The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191738.1 | NP_001178667.1 | 4 - 5 | 1933 | 60 |
XM_006234311.1 | XP_006234373.1 | 4 - 5 | 742 | 60 |